Anti-MYBPC3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19977-1-AP-150UL
10090-676EA 537.7 USD
10090-676
Anti-MYBPC3 Rabbit Polyclonal Antibody
Antibodies
MYBPC3 belongs to the immunoglobulin superfamily and MyBP family. MYBPC3 located in the crossbridge region of vertebrate striated muscle a bands. In vitro it binds MHC, F-actin and native thin filaments, and modifies the activity of actin-activated myosin ATPase. It may modulate muscle contraction or may play a more structural role. Defects in MYBPC3 are the cause of cardiomyopathy familial hypertrophic type 4 (CMH4). The antibody recognizes the C-term of MYBPC3.

Western Blot: Human Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: MYBPC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR