Anti-LHCGR Rabbit Polyclonal Antibody

Supplier: Proteintech
19968-1-AP-150UL
10089-622EA 524.34 USD
10089-622
Anti-LHCGR Rabbit Polyclonal Antibody
Antibodies
LHCGR, also named as LCGR, LGR2, LHRHR LHR and LSHR, belongs to the G-protein coupled receptor 1 family and FSH/LSH/TSH subfamily. LHCGR is a receptor for lutropin-choriogonadotropic hormone. The activity of LHCGR is mediated by G proteins which activate adenylate cyclase. Defects in LHCGR are a cause of familial male precocious puberty (FMPP). Defects in LHCGR are a cause of Leydig cell hypoplasia (LCH).

Western Blot: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: LHCGR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR