Anti-KCNB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19963-1-AP-150UL
10089-124EA 402 USD
10089-124
Anti-KCNB1 Rabbit Polyclonal Antibody
Antibodies
KCNB1, also named as DRK1, KV2.1, and h-DRK1, belongs to the potassium channel family and B (Shab) subfamily. KCNB1 mediates the voltage-dependent potassium ion permeability of excitable membranes. Channels open or close in response to the voltage difference across the membrane, letting potassium ions pass in accordance with their electrochemical gradient. The antibody is specific to KCNB1.

IF:SH-SY5Y Cells, 1:10-1:100

Type: Primary
Antigen: KCNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR