Anti-ITPR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19962-1-AP-150UL
10089-052EA 402 USD
10089-052
Anti-ITPR1 Rabbit Polyclonal Antibody
Antibodies
ITPR1, also named as INSP3R1 and IP3R, belongs to the InsP3 receptor family. It is a receptor for inositol 1,4,5-trisphosphate which is a second messenger that mediates the release of intracellular calcium. Defects in ITPR1 are the cause of spinocerebellar ataxia type 15 (SCA15). The antibody has no cross reaction with ITPR2 and ITPR3.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: ITPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR