Anti-GAC Rabbit Polyclonal Antibody

Supplier: Proteintech
19958-1-AP-150UL
10087-242EA 402 USD
10087-242
Anti-GAC Rabbit Polyclonal Antibody
Antibodies
GAC is isoform GAC of GLS which also named as GLS1, KIAA0838, L-glutamine amidohydrolase and K-glutaminase. GLS belongs to the glutaminase family. GLS catalyzes the first reaction in the primary pathway for the renal catabolism of glutamine. The reaction is: L-glutamine + H2O = L-glutamate + NH3.GAC is dysregulated in HIV-1-infected microglia. This antibody is specific to GAC isoform. It has no cross reaction to KGA.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200; IP: Mouse Kidney Tissue, 1:200-1:2000

Type: Primary
Antigen: GAC-specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR