Anti-MGLUR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19956-1-AP-150UL
10090-194EA 402 USD
10090-194
Anti-MGLUR2 Rabbit Polyclonal Antibody
Antibodies
GRM2, also named as GPRC1B and MGLUR2, belongs to the G-protein coupled receptor 3 family. GRM2 is a receptor for glutamate. The activity of GRM2 is mediated by a G-protein that inhibits adenylate cyclase activity. It may mediate suppression of neurotransmission or may be involved in synaptogenesis or synaptic stabilization.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: MGLUR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR