Anti-GPR22 Rabbit Polyclonal Antibody

Supplier: Proteintech
19951-1-AP-150UL
10087-790EA 402 USD
10087-790
Anti-GPR22 Rabbit Polyclonal Antibody
Antibodies
GPR22 Belongs to the G-protein coupled receptor 1 family. GPR22 is an Orphan receptor. The antibody recognize the C-term of GPR22.

Western Blot: Mouse Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: GPR22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR