Anti-ERBB4 Rabbit Polyclonal Antibody

Supplier: Proteintech
19943-1-AP-150UL
10086-604EA 402 USD
10086-604
Anti-ERBB4 Rabbit Polyclonal Antibody
Antibodies
ERBB4, also named as p180erbB4 and HER4, belongs to the protein kinase superfamily, Tyr protein kinase family and EGF receptor subfamily. ERBB4 specifically binds and it is activated by neuregulins, NRG-2, NRG-3, heparin-binding EGF-like growth factor, betacellulin and NTAK. Interaction with these factors induces cell differentiation. ERBB4 is not activated by EGF, TGF-A, and amphiregulin. ERBB4 catalyze the reaction: ATP + a [protein]-L-tyrosine = ADP + a [protein]-L-tyrosine phosphate. The antibody recognize the C-term of ERBB4.

Western Blot: Mouse Brain Tissue, 1:200-1:1000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: ERBB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR