Anti-CYP2E1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19937-1-AP-150UL
10085-522EA 402 USD
10085-522
Anti-CYP2E1 Rabbit Polyclonal Antibody
Antibodies
CYP2E1, also named as CYPIIE1, 4-nitrophenol 2-hydroxylase, P450-J and CYP2E, belongs to the cytochrome P450 family. CYP2E1 metabolizes several precarcinogens, drugs, and solvents to reactive metabolites. It inactivates a number of drugs and xenobiotics and also bioactivates many xenobiotic substrates to their hepatotoxic or carcinogenic forms. CYP2E1 catalyze the reaction: 4-nitrophenol + NADPH + O2 = 4-nitrocatechol + NADP+ + H2O. The antibody is specific to CYP2E1.

Western Blot:Jurkat Cells, 1:200-1:2000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: CYP2E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR