Anti-HEY1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19929-1-AP-150UL
10087-296EA 402 USD
10087-296
Anti-HEY1 Rabbit Polyclonal Antibody
Antibodies
Hairy/enhancer of split-related proteins are basic helix-loop-helix (bHLH) transcription factors implicated in cell fate decision and boundary formation. And HEY1 is one of such protein. It is the direct downstream effector of Notch signaling which may be required for cardiovascular development. It preferentially bind to the canonical E box sequence 5'-CACGTG-3' and acts as a transcriptional repressor. HEY1 is a 33kDa protein and forms dimer by self-associating.

Western Blot:DU 145 Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: HEY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR