Anti-C12ORF11 Rabbit Polyclonal Antibody

Supplier: Proteintech
19892-1-AP-150UL
10083-758EA 402 USD
10083-758
Anti-C12ORF11 Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF7 Cells, 1:1000-1:10000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: C12ORF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR