Anti-EGLN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19886-1-AP-150UL
10081-358EA 402 USD
10081-358
Anti-EGLN1 Rabbit Polyclonal Antibody
Antibodies
EGLN1(Egl nine homolog 1) is also named as HIF-PH2,HPH-2,PHD2.It is the most important isozyme under normoxia and, through regulating the stability of HIF1, involved in various hypoxia-influenced processes such as angiogenesis in retinal and cardiac functionality. EGLN1 plays a critical role in regulating HIF levels in EPO-producing cells in humans.Defects in EGLN1 are the cause of familial erythrocytosis type 3 (ECYT3).This antibody is specific to EGLN1.

Western Blot: Mouse Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: EGLN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR