Anti-Spermidine synthase Rabbit Polyclonal Antibody

Supplier: Proteintech
19858-1-AP-150UL
10095-166EA 402 USD
10095-166
Anti-Spermidine synthase Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: Spermidine synthase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR