Anti-TMEM173 Rabbit Polyclonal Antibody

Supplier: Proteintech
19851-1-AP-150UL
10096-092EA 537.7 USD
10096-092
Anti-TMEM173 Rabbit Polyclonal Antibody
Antibodies
TMEM173, also named as ERIS, MITA, STING and MPYS, is a facilitator of innate immune signaling that promotes the production of type I interferon (IFN-alpha and IFN-beta). TMEM173 mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. There’re monomer (40 kDa) and dimer (80 kDa) form of TMEM173 in western blot test.

Western Blot: Mouse Thymus Tissue, 1:500-1:5000; IHC: Human spleen Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: TMEM173
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR