Anti-STAG2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19837-1-AP-150UL
10095-236EA 402 USD
10095-236
Anti-STAG2 Rabbit Polyclonal Antibody
Antibodies
STAG2, also named as SA2, is a component of cohesin complex, a complex required for the cohesion of sister chromatids after DNA replication. The cohesin complex may play a role in spindle pole assembly during mitosis. Mutations in STAG2 appear to be a first step in the transformation of a normal cell into a cancer cell.

Western Blot:Jurkat Cells, 1:1000-1:10000; IHC: Human cervical cancer, 1:20-1:200; IF:293, 1:10-1:100

Type: Primary
Antigen: STAG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR