Anti-TM7SF3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19826-1-AP-150UL
10096-056EA 402 USD
10096-056
Anti-TM7SF3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Kidney Tissue, 1:1000-1:10000

Type: Primary
Antigen: TM7SF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR