Anti-TLR4 Rabbit Polyclonal Antibody

Supplier: Proteintech
19811-1-AP-150UL
10096-046EA 537.7 USD
10096-046
Anti-TLR4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:rat Liver Tissue, N/A

Type: Primary
Antigen: TLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR