Anti-TLR5 Rabbit Polyclonal Antibody

Supplier: Proteintech
19810-1-AP-150UL
10096-048EA 524.48 USD
10096-048
Anti-TLR5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A549 Cells, 1:200-1:2000

Type: Primary
Antigen: TLR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR