Anti-Collagen Type XVI Rabbit Polyclonal Antibody

Supplier: Proteintech
19802-1-AP-150UL
10084-980EA 402 USD
10084-980
Anti-Collagen Type XVI Rabbit Polyclonal Antibody
Antibodies
COL16A1, also named as FP1572, belongs to the fibril-associated collagens with interrupted helices (FACIT) family. COL16A1 is involved in mediating cell attachment and inducing integrin-mediated cellular reactions, such as cell spreading and alterations in cell morphology. The antibody is specific to COL16A1.

Western Blot: Human Placenta Tissue, 1:500-1:5000

Type: Primary
Antigen: Collagen Type XVI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR