Anti-ADGRB3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19789-1-AP-150UL
10083-494EA 402 USD
10083-494
Anti-ADGRB3 Rabbit Polyclonal Antibody
Antibodies
BAI3, also named as KIAA0550, belongs to the G-protein coupled receptor 2 family and LN-TM7 subfamily. BAI3 might be involved in angiogenesis inhibition and suppression of glioblastoma. The antibody is special to BAI3.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: ADGRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR