Anti-APBA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19781-1-AP-150UL
10082-662EA 402 USD
10082-662
Anti-APBA2 Rabbit Polyclonal Antibody
Antibodies
APBA2, also named as MINT2 and X11L, is putative function in synaptic vesicle exocytosis by binding to STXBP1, an essential component of the synaptic vesicle exocytotic machinery. APBA2 may modulate processing of the beta-amyloid precursor protein (APP) and hence formation of beta-APP.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: APBA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR