Anti-ADRA2B Rabbit Polyclonal Antibody

Supplier: Proteintech
19778-1-AP-150UL
10082-548EA 402 USD
10082-548
Anti-ADRA2B Rabbit Polyclonal Antibody
Antibodies
ADRA2B, also named as ADRA2L1 and ADRA2RL1, belongs to the G-protein coupled receptor 1 family. ADRA2B mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G proteins. The antibody is specific to ADRA2B.

Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: ADRA2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR