Anti-ADRA1A Rabbit Polyclonal Antibody

Supplier: Proteintech
19777-1-AP-150UL
10082-544EA 402 USD
10082-544
Anti-ADRA1A Rabbit Polyclonal Antibody
Antibodies
ADRA1A, also named as ADRA1C, belongs to the G-protein coupled receptor 1 family. ADRA1A mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system. Its effect is mediated by G(q) and G(11) proteins. The antibody is specific to ADRA1A.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: ADRA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR