Anti-Ryanodine Receptor 2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19765-1-AP-150UL
10093-746EA 524.48 USD
10093-746
Anti-Ryanodine Receptor 2 Rabbit Polyclonal Antibody
Antibodies
RYR2 belongs to the ryanodine receptor family. RYR2 provides communication between transverse-tubules and sarcoplasmic reticulum. Contraction of cardiac muscle is triggered by release of calcium ions from SR following depolarization of T-tubules. Defects in RYR2 are the cause of familial arrhythmogenic right ventricular dysplasia type 2 (ARVD2) which known as arrhythmogenic right ventricular cardiomyopathy 2 (ARVC2). Defects in RYR2 are the cause of catecholaminergic polymorphic ventricular tachycardia type 1 (CPVT1) which known as stress-induced polymorphic ventricular tachycardia (VTSIP).

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: Ryanodine Receptor 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR