Anti-LATH Rabbit Polyclonal Antibody

Supplier: Proteintech
19763-1-AP-150UL
10089-532EA 402 USD
10089-532
Anti-LATH Rabbit Polyclonal Antibody
Antibodies
LATH, also named as BASE, belongs to the BPI/LBP/Plunc superfamily and Plunc family. It is the major protein in sweat, has surfactant properties.

IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: LATH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR