Anti-SFT2D3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19758-1-AP-150UL
10094-468EA 524.48 USD
10094-468
Anti-SFT2D3 Rabbit Polyclonal Antibody
Antibodies
SFT2D3 belongs to the SFT2 family. SFT2D3 may be involved in fusion of retrograde transport vesicles derived from an endocytic compartment with the Golgi complex.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: SFT2D3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR