Anti-WASF4 Rabbit Polyclonal Antibody

Supplier: Proteintech
19755-1-AP-150UL
10097-066EA 402 USD
10097-066
Anti-WASF4 Rabbit Polyclonal Antibody
Antibodies
WASF4, also named as SCAR2, belongs to the SCAR/WAVE family. WASF4 is a downstream effector molecule which involved in the transmission of signals from tyrosine kinase receptors and small GTPases to the actin cytoskeleton. The antibody is specific to WASF4.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: WASF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR