Anti-BTF3L3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19753-1-AP-150UL
10083-714EA 402 USD
10083-714
Anti-BTF3L3 Rabbit Polyclonal Antibody
Antibodies
BTF3L3 belongs to the NAC-beta family. It is a general transcription factor. It is required for the initiation of transcription. The antibody has no cross reaction to BTF3.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: BTF3L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR