Anti-FOXJ3 Rabbit Polyclonal Antibody

Supplier: Proteintech
19751-1-AP-150UL
10081-782EA 402 USD
10081-782
Anti-FOXJ3 Rabbit Polyclonal Antibody
Antibodies
FOXJ3, also named as KIAA1041, contains 1 fork-head DNA-binding domain. FOXJ3 is a transcriptional activator.

Western Blot: Human Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: FOXJ3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR