Anti-ARSK Rabbit Polyclonal Antibody

Supplier: Proteintech
19749-1-AP-150UL
10083-168EA 402 USD
10083-168
Anti-ARSK Rabbit Polyclonal Antibody
Antibodies
ARSK, also named as TSULF and ASK, belongs to the sulfatase family. ARSK binds 1 calcium ion per subunit. The antibody is specific to ARSK.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: ARSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR