Anti-SLK Rabbit Polyclonal Antibody

Supplier: Proteintech
19743-1-AP-150UL
10094-824EA 402 USD
10094-824
Anti-SLK Rabbit Polyclonal Antibody
Antibodies
SLK, also named as KIAA0204 and STK2, belongs to the protein kinase superfamily, STE Ser/Thr protein kinase family and STE20 subfamily. SLK mediates apoptosis and actin stress fiber dissolution. The antibody recognizes the C-term of SLK.

Western Blot: HEK-293 Cells, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: SLK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR