Anti-UCN Rabbit Polyclonal Antibody

Supplier: Proteintech
19731-1-AP-150UL
10096-814EA 402 USD
10096-814
Anti-UCN Rabbit Polyclonal Antibody
Antibodies
Urocortin is a member of the sauvagine/corticotropin-releasing factor/urotensin I family that includes CRF, urotensin I, sauvagine, urocortin II and urocortin III. . Urocortin is a potent anorexigenic peptide that induces fed-like motor activity when administered centrally or peripherally in fasted animals. Urocortin is also a potent and long-lasting hypotensive agent and increases coronary blood flow. Urocortin acts in vitro to stimulate the secretion of adrenocorticotropic hormone (ACTH) and binds with high affinity to CRF receptor types 1, 2-alpha, and 2-beta. Urocortin is synthesized as precursor, which is subsequently processed to the mature biologically active peptide (a 40-amino acid Molecule ).

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: UCN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR