Anti-TMF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19728-1-AP-150UL
10095-884EA 402 USD
10095-884
Anti-TMF1 Rabbit Polyclonal Antibody
Antibodies
TMF1, also named as ARA160, is a potential coactivator of the androgen receptor. TMF1 mediates STAT3 degradation. TMF1 plays critical roles in two RAB6-dependent retrograde transport processes: one from endosomes to the Golgi and the other from the Golgi to the ER. TMF1 binds the HIV-1 TATA element and inhibits transcriptional activation by the TATA-binding protein (TBP). The antibody is specific to TMF1.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: TMF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR