Anti-Thrombospondin 3-Specific Rabbit Polyclonal Antibody

Supplier: Proteintech
19727-1-AP-150UL
10095-442EA 402 USD
10095-442
Anti-Thrombospondin 3-Specific Rabbit Polyclonal Antibody
Antibodies
THBS3, also named as TSP3, belongs to the thrombospondin family. THBS3 is an adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. THBS3 can bind to fibrinogen, fibronectin, laminin and type V collagen. The antibody is specific to THBS3.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000

Type: Primary
Antigen: Thrombospondin 3-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR