Anti-STATH Rabbit Polyclonal Antibody

Supplier: Proteintech
19724-1-AP-150UL
10095-270EA 402 USD
10095-270
Anti-STATH Rabbit Polyclonal Antibody
Antibodies
STATH belongs to the histatin/statherin family. STATH is a salivary protein that stabilizes saliva supersaturated with calcium salts by inhibiting the precipitation of calcium phosphate salts. It also modulates hydroxyapatite crystal formation on the tooth surface.

Western Blot: Mouse pancreas Tissue, 1:500-1:5000

Type: Primary
Antigen: STATH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR