Anti-PYGM Rabbit Polyclonal Antibody

Supplier: Proteintech
19716-1-AP-150UL
10093-114EA 402 USD
10093-114
Anti-PYGM Rabbit Polyclonal Antibody
Antibodies
PYGM belongs to the glycogen phosphorylase family. Phosphorylase is an important allosteric enzyme in carbohydrate metabolism. Enzymes from different sources differ in their regulatory mechanisms and in their natural substrates. However, all known phosphorylases share catalytic and structural properties. Defects in PYGM are the cause of glycogen storage disease type 5 (GSD5) which also known as McArdle disease. The antibody is specific to PYGM.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Skeletal Muscle, 1:20-1:200

Type: Primary
Antigen: PYGM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR