Anti-PTPRCAP / CD45AP Rabbit Polyclonal Antibody

Supplier: Proteintech
19715-1-AP-150UL
10093-074EA 402 USD
10093-074
Anti-PTPRCAP / CD45AP Rabbit Polyclonal Antibody
Antibodies
PTPRCAP, also named as LPAP and CD45-AP, interacts with CD45/PTPRC.

Western Blot:Jurkat Cells, 1:200-1:1000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: PTPRCAP / CD45AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR