Anti-NEB-Specific Rabbit Polyclonal Antibody

Supplier: Proteintech
19706-1-AP-150UL
10090-996EA 402 USD
10090-996
Anti-NEB-Specific Rabbit Polyclonal Antibody
Antibodies
NEB, a giant muscle protein, may be involved in maintaining the structural integrity of sarcomeres and the membrane system associated with the myofibrils. NEB binds and stabilizes F-actin. Defects in NEB are the cause of nemaline myopathy type 2 (NEM2). The antibody is specific to NEB.

IHC: Human Skeletal Muscle, 1:20-1:200

Type: Primary
Antigen: NEB-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR