Anti-ND1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19703-1-AP-150UL
10090-890EA 537.7 USD
10090-890
Anti-ND1 Rabbit Polyclonal Antibody
Antibodies
ND1 belongs to the complex I subunit 1 family. ND1 is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR