Anti-LRP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19700-1-AP-150UL
10089-790EA 402 USD
10089-790
Anti-LRP2 Rabbit Polyclonal Antibody
Antibodies
LRP2, also named as gp330 and Megalin, belongs to the LDLR family. It acts together with cubilin to mediate HDL endocytosis. LRP2 may participate in regulation of parathyroid-hormone and para-thyroid-hormone-related protein release. Defects in LRP2 are the cause of Donnai-Barrow syndrome (DBS). The antibody is specific to LRP2.

Western Blot: HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: LRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR