Anti-LAMC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19698-1-AP-150UL
10089-482EA 402 USD
10089-482
Anti-LAMC2 Rabbit Polyclonal Antibody
Antibodies
LAMC2, also named as LAMB2T and LAMNB2, is thought to mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. LAMC2 binds to cells via a high affinity receptor. Laminin exerts cell-scattering activity toward a wide variety of cells, including epithelial, endothelial, and fibroblastic cells.

Western Blot:A431 Cells, 1:500-1:5000

Type: Primary
Antigen: LAMC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR