Anti-Kv1.4-Specific Rabbit Polyclonal Antibody

Supplier: Proteintech
19697-1-AP-150UL
10088-752EA 402 USD
10088-752
Anti-Kv1.4-Specific Rabbit Polyclonal Antibody
Antibodies
KCNA4, also named as KCNA4L, HBK4, HUKII, HK1 and HPCN2, belongs to the potassium channel family and A (Shaker) subfamily. KCN4 mediates the voltage-dependent potassium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a potassium-selective channel through which potassium ions may pass in accordance with their electrochemical gradient. The antibody is specific to KCN4.

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: Kv1.4-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR