Anti-Integrin beta-6-Specific Rabbit Polyclonal Antibody

Supplier: Proteintech
19695-1-AP-150UL
10089-030EA 402 USD
10089-030
Anti-Integrin beta-6-Specific Rabbit Polyclonal Antibody
Antibodies
ITGB6 belongs to the integrin beta chain family. ITGB6 is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalisation of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion. The antibody is specific to ITGB6.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human lung Tissue, 1:20-1:200

Type: Primary
Antigen: Integrin beta-6-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR