Anti-BLOC1S1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19687-1-AP-150UL
10083-630EA 402 USD
10083-630
Anti-BLOC1S1 Rabbit Polyclonal Antibody
Antibodies
BLOC1S1, also named as BLOS1, GCN5L1 and RT14, belongs to the BLOC1S1 family. BLOC1S1 may be involved in the biogenesis of specialized organelles of the endosomal-lysosomal system. The antibody is specific to NLOC1S1.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human small intestine Tissue, 1:20-1:200

Type: Primary
Antigen: BLOC1S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR