Anti-CACNA1B Rabbit Polyclonal Antibody

Supplier: Proteintech
19681-1-AP-150UL
10083-934EA 402 USD
10083-934
Anti-CACNA1B Rabbit Polyclonal Antibody
Antibodies
CACNA1B, also named as CACH5, CACNL1A5 and BIII, belongs to the calcium channel alpha-1 subunit (TC 1.A.1.11) family. Voltage-sensitive calcium channels (VSCC) mediate the entry of calcium ions into excitable cells and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, gene expression, cell motility, cell division and cell death. CACNA1B gives rise to N-type calcium currents. N-type calcium channels belong to the 'high-voltage activated' (HVA) group and are blocked by omega-conotoxin-GVIA (omega-CTx-GVIA) and by omega-agatoxin-IIIA (omega-Aga-IIIA). They are however insensitive to dihydropyridines (DHP), and omega-agatoxin-IVA (omega-Aga-IVA). CACNA1B may play a role in directed migration of immature neurons.

Western Blot: Mouse Brain Tissue, 1:200-1:1000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: CACNA1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR