Anti-HSPG2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19675-1-AP-150UL
10087-636EA 402 USD
10087-636
Anti-HSPG2 Rabbit Polyclonal Antibody
Antibodies
HSPG2, also named as PLC and HSPG, is an integral component of basement membranes. It is responsible for the fixed negative electrostatic charge and is involved in the charge-selective ultrafiltration properties. It serves as an attachment substrate for cells. Defects in HSPG2 are the cause of Schwartz-Jampel syndrome (SJS1). Defects in HSPG2 are the cause of dyssegmental dysplasia Silverman-Handmaker type (DDSH). The antibody is specific to HSPG2.

Western Blot:COLO 320 Cells, 1:200-1:1000

Type: Primary
Antigen: HSPG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR