Anti-MUTYH Rabbit Polyclonal Antibody

Supplier: Proteintech
19650-1-AP-150UL
10090-662EA 402 USD
10090-662
Anti-MUTYH Rabbit Polyclonal Antibody
Antibodies
MUTYH(MutY homolog) is also named as MYH and belongs to the Nth/MutY family. The gene enconde an A/G-specific adenine DNA glycosylase that it is involved in oxidative DNA damage repair and initiates repair of A*oxoG to C*G by removing the inappropriately paired adenine base from the DNA backbone. Possesses both adenine and 2-OH-A DNA glycosylase activities. Defects in MUTYH are a cause of familial adenomatous polyposis type 2 (FAP2) and gastric cancer (GASC). It has 6 isoforms produced by alternative splicing with the molecular weight between 57 kDa and 60 kDa.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: MUTYH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR