Anti-TRIP13 Rabbit Polyclonal Antibody

Supplier: Proteintech
19602-1-AP-150UL
10096-368EA 402 USD
10096-368
Anti-TRIP13 Rabbit Polyclonal Antibody
Antibodies
TRIP13 gene encodes thyroid receptor-interacting protein 13, that interact with thyroid hormone receptors, also known as human papillomavirus type 16 E1 protein-binding protein (16E1BP). This protein belongs to the AAA ATPase family and the PCH2 subfamily. TRIP13 gene may be one of the several playing roles in early-stage non-small cell lung cancer. TRIP13 also functions in chromosome recombination and chromosome structure development during meiosis, and it is required for development of higher-order chromosome structures and is needed for synaptonemal-complex formation.

Western Blot:Jurkat Cells, 1:500-1:5000; IP: Mouse Testis Tissue, 1:200-1:1000

Type: Primary
Antigen: TRIP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR