Anti-PGPEP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19566-1-AP-150UL
10092-162EA 402 USD
10092-162
Anti-PGPEP1 Rabbit Polyclonal Antibody
Antibodies
PGPEP1, also named as PGPI, belongs to the peptidase C15 family. PGPEP1 removes 5-oxoproline from various penultimate amino acid residues except L-proline.

Western Blot: Human Brain Tissue, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: PGPEP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR