Anti-SLC12A5 Rabbit Polyclonal Antibody

Supplier: Proteintech
19565-1-AP-150UL
10094-698EA 402 USD
10094-698
Anti-SLC12A5 Rabbit Polyclonal Antibody
Antibodies
SLC12A5, also named as KCC2 and KIAA1176, belongs to the SLC12A transporter family. SLC12A5 mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. SLC12A5 is important for Cl- homeostasis in neurons. The antibody is specific to SLC12A5.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: SLC12A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR